Assay Details
| Gene Symbol: LOC4331860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776986.2 | XP_015632472.1 | 6 - 7 | 1205 | 70 | |
| XM_015776987.2 | XP_015632473.1 | 6 - 7 | 1192 | 70 | ||
| GenBank mRNA | AK242821.1 | - | 5 - 6 | 868 | 70 | |
More Information
Set Membership: |
|

LOC4331860
Genomic Map