Assay Details
| Gene Symbol: LOC4332041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772537.2 | XP_015628023.1 | 9 - 10 | 1293 | 71 | |
| GenBank mRNA | AK119546.1 | - | 9 - 10 | 1211 | 71 | |
| AY129294.1 | - | 9 - 10 | 1293 | 71 | ||
More Information
Set Membership: |
|

LOC4332041
Genomic Map