Assay Details
| Gene Symbol: LOC4332578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773015.2 | XP_015628501.1 | 17 - 18 | 2652 | 87 | |
| GenBank mRNA | AK106465.1 | - | 5 - 6 | 907 | 87 | |
More Information
Set Membership: |
|

LOC4332578
Genomic Map