Assay Details
| Gene Symbol: LOC4332578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773015.2 | XP_015628501.1 | 18 - 19 | 3147 | 106 | |
| GenBank mRNA | AK106465.1 | - | 6 - 7 | 1402 | 106 | |
More Information
Set Membership: |
|

LOC4332578
Genomic Map