Assay Details
| Gene Symbol: LOC4332593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775960.2 | XP_015631446.1 | 1 - 2 | 346 | 111 | |
| GenBank mRNA | AK101765.1 | - | 1 - 2 | 344 | 111 | |
More Information
Set Membership: |
|

LOC4332593
Genomic Map