Assay Details
| Gene Symbol: LOC4332593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775960.2 | XP_015631446.1 | 2 - 3 | 930 | 65 | |
| GenBank mRNA | AK101765.1 | - | 2 - 3 | 928 | 65 | |
More Information
Set Membership: |
|

LOC4332593
Genomic Map