Assay Details
| Gene Symbol: LOC4332615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772777.2 | XP_015628263.1 | 1 - 2 | 250 | 111 | |
| GenBank mRNA | AB116073.1 | - | 1 - 2 | 199 | 111 | |
| AK060518.1 | - | 1 - 2 | 154 | 111 | ||
| AK099107.1 | - | 1 - 2 | 132 | 111 | ||
| AK105977.1 | - | 1 - 2 | 197 | 111 | ||
More Information
Set Membership: |
|

LOC4332615
Genomic Map