Assay Details
| Gene Symbol: LOC4332627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776835.2 | XP_015632321.1 | 1 - 2 | 434 | 70 | |
| GenBank mRNA | AK060625.1 | - | 1 - 2 | 434 | 70 | |
More Information
Set Membership: |
|

LOC4332627
Genomic Map