Assay Details
| Gene Symbol: LOC4333632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777194.2 | XP_015632680.1 | 2 - 3 | 825 | 58 | |
| GenBank mRNA | AB104733.1 | - | 2 - 3 | 785 | 58 | |
| AK121321.1 | - | 2 - 3 | 786 | 58 | ||
More Information
Set Membership: |
|

LOC4333632
Genomic Map