Assay Details
| Gene Symbol: LOC4333842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774844.2 | XP_015630330.1 | 3 - 4 | 508 | 138 | |
| GenBank mRNA | AK120790.1 | - | 3 - 4 | 481 | 138 | |
More Information
Set Membership: |
|

LOC4333842
Genomic Map