Assay Details
| Gene Symbol: LOC4333842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774844.2 | XP_015630330.1 | 6 - 7 | 909 | 123 | |
| GenBank mRNA | AK105850.1 | - | 2 - 3 | 265 | 123 | |
| AK120790.1 | - | 6 - 7 | 882 | 123 | ||
More Information
Set Membership: |
|

LOC4333842
Genomic Map