Assay Details
| Gene Symbol: LOC4333842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774844.2 | XP_015630330.1 | 8 - 9 | 1246 | 69 | |
| GenBank mRNA | AK059517.1 | - | 3 - 4 | 473 | 69 | |
| AK105850.1 | - | 4 - 5 | 602 | 69 | ||
| AK120790.1 | - | 8 - 9 | 1219 | 69 | ||
More Information
Set Membership: |
|

LOC4333842
Genomic Map