Assay Details
| Gene Symbol: LOC4333842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774844.2 | XP_015630330.1 | 9 - 10 | 1284 | 102 | |
| GenBank mRNA | AK059517.1 | - | 4 - 5 | 511 | 102 | |
| AK105850.1 | - | 5 - 6 | 640 | 102 | ||
| AK120790.1 | - | 9 - 10 | 1257 | 102 | ||
More Information
Set Membership: |
|

LOC4333842
Genomic Map