Assay Details
| Gene Symbol: LOC4333963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772462.2 | XP_015627948.1 | 4 - 5 | 852 | 149 | |
| GenBank mRNA | AK063969.1 | - | 4 - 5 | 454 | 149 | |
| AK073636.1 | - | 4 - 5 | 567 | 149 | ||
More Information
Set Membership: |
|

LOC4333963
Genomic Map