Assay Details
| Gene Symbol: LOC4334049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774323.2 | XP_015629809.1 | 3 - 4 | 830 | 72 | |
| GenBank mRNA | AK073529.1 | - | 3 - 4 | 830 | 72 | |
| X64396.1 | - | 3 - 4 | 812 | 72 | ||
More Information
Set Membership: |
|

LOC4334049
Genomic Map