Assay Details
| Gene Symbol: LOC4334101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777569.2 | XP_015633055.1 | 2 - 3 | 262 | 88 | |
| XM_015777570.2 | XP_015633056.1 | 2 - 3 | 203 | 88 | ||
| GenBank mRNA | AF073697.1 | - | 2 - 3 | 179 | 88 | |
| AK071279.1 | - | 2 - 3 | 154 | 88 | ||
| BT129303.1 | - | 2 - 3 | 136 | 88 | ||
More Information
Set Membership: |
|

LOC4334101
Genomic Map