Assay Details
| Gene Symbol: LOC9267055 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775038.2 | XP_015630524.1 | 3 - 4 | 336 | 89 | |
| XM_015775039.2 | XP_015630525.1 | 3 - 4 | 337 | 89 | ||
| GenBank mRNA | AB219537.1 | - | 3 - 4 | 292 | 89 | |
| AK287862.1 | - | 3 - 4 | 340 | 89 | ||
| GU170363.1 | - | 3 - 4 | 292 | 89 | ||
More Information
Set Membership: |
|

LOC9267055
Genomic Map