Assay Details
| Gene Symbol: LOC4334171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775934.2 | XP_015631420.1 | 7 - 8 | 1472 | 152 | |
| GenBank mRNA | AK063766.1 | - | 6 - 7 | 1123 | 152 | |
| KT199368.1 | - | 2 - 3 | 239 | 152 | ||
More Information
Set Membership: |
|

LOC4334171
Genomic Map