Assay Details
| Gene Symbol: LOC4337272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779836.2 | XP_015635322.1 | 12 - 13 | 1304 | 61 | |
| GenBank mRNA | AK065491.1 | - | 12 - 13 | 1162 | 61 | |
| AK099252.1 | - | 12 - 13 | 1134 | 61 | ||
| BT131774.1 | - | 1 - 2 | 37 | 61 | ||
More Information
Set Membership: |
|

LOC4337272
Genomic Map