Assay Details
| Gene Symbol: LOC4337272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779836.2 | XP_015635322.1 | 1 - 2 | 183 | 79 | |
| GenBank mRNA | AK063706.1 | - | 1 - 2 | 41 | 79 | |
| AK065491.1 | - | 1 - 2 | 42 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015779836.2 transcripts.
Set Membership: |
|

LOC4337272
Genomic Map