Assay Details
| Gene Symbol: LOC4337272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779836.2 | XP_015635322.1 | 2 - 3 | 464 | 73 | |
| GenBank mRNA | AK063706.1 | - | 2 - 3 | 322 | 73 | |
| AK065491.1 | - | 2 - 3 | 323 | 73 | ||
| AK099252.1 | - | 2 - 3 | 294 | 73 | ||
More Information
Set Membership: |
|

LOC4337272
Genomic Map