Assay Details
| Gene Symbol: LOC4339002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784509.2 | XP_015639995.1 | 4 - 5 | 747 | 85 | |
| GenBank mRNA | AK069936.1 | - | 4 - 5 | 721 | 85 | |
More Information
Set Membership: |
|

LOC4339002
Genomic Map