Assay Details
| Gene Symbol: LOC4339026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784239.2 | XP_015639725.1 | 1 - 2 | 259 | 91 | |
| XM_015784240.2 | XP_015639726.1 | 1 - 2 | 259 | 91 | ||
| GenBank mRNA | AK099608.1 | - | 1 - 2 | 207 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784239.2, XM_015784240.2 transcripts.
Set Membership: |
|

LOC4339026
Genomic Map