Assay Details
| Gene Symbol: LOC4339026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784239.2 | XP_015639725.1 | 2 - 3 | 2340 | 89 | |
| XM_015784240.2 | XP_015639726.1 | 3 - 4 | 2040 | 89 | ||
| GenBank mRNA | AK099608.1 | - | 2 - 3 | 2288 | 89 | |
More Information
Set Membership: |
|

LOC4339026
Genomic Map