Assay Details
| Gene Symbol: LOC4340102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788795.2 | XP_015644281.1 | 5 - 6 | 1397 | 96 | |
| GenBank mRNA | AK103630.1 | - | 5 - 6 | 1373 | 96 | |
More Information
Set Membership: |
|

LOC4340102
Genomic Map