Assay Details
| Gene Symbol: LOC4340102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788795.2 | XP_015644281.1 | 6 - 7 | 1507 | 82 | |
| GenBank mRNA | AK103630.1 | - | 6 - 7 | 1483 | 82 | |
More Information
Set Membership: |
|

LOC4340102
Genomic Map