Assay Details
| Gene Symbol: LOC4340240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787738.2 | XP_015643224.1 | 1 - 2 | 244 | 71 | |
| GenBank mRNA | AK065744.1 | - | 1 - 2 | 194 | 71 | |
More Information
Set Membership: |
|

LOC4340240
Genomic Map