Assay Details
| Gene Symbol: LOC4340283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785911.2 | XP_015641397.1 | 3 - 4 | 494 | 97 | |
| XM_015785912.2 | XP_015641398.1 | 3 - 4 | 494 | 97 | ||
| GenBank mRNA | AK119521.1 | - | 3 - 4 | 487 | 97 | |
More Information
Set Membership: |
|

LOC4340283
Genomic Map