Assay Details
| Gene Symbol: LOC4340341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787450.2 | XP_015642936.1 | 2 - 3 | 448 | 134 | |
| GenBank mRNA | AK243433.1 | - | 2 - 3 | 390 | 134 | |
More Information
Set Membership: |
|

LOC4340341
Genomic Map