Assay Details
| Gene Symbol: LOC4340408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785715.2 | XP_015641201.1 | 1 - 2 | 156 | 105 | |
| GenBank mRNA | AK060939.1 | - | 1 - 2 | 110 | 105 | |
| AK064613.1 | - | 1 - 2 | 113 | 105 | ||
| AK100138.1 | - | 1 - 2 | 150 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015785715.2 transcripts.
Set Membership: |
|

LOC4340408
Genomic Map