Assay Details
| Gene Symbol: LOC4340443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788564.2 | XP_015644050.1 | 7 - 8 | 1247 | 142 | |
| XM_015788566.2 | XP_015644052.1 | 5 - 6 | 1092 | 142 | ||
| GenBank mRNA | AK241861.1 | - | 5 - 6 | 997 | 142 | |
More Information
Set Membership: |
|

LOC4340443
Genomic Map