Assay Details
| Gene Symbol: LOC4340443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788564.2 | XP_015644050.1 | 10 - 11 | 1885 | 79 | |
| XM_015788566.2 | XP_015644052.1 | 8 - 9 | 1730 | 79 | ||
| GenBank mRNA | AK241861.1 | - | 8 - 9 | 1635 | 79 | |
More Information
Set Membership: |
|

LOC4340443
Genomic Map