Assay Details
| Gene Symbol: LOC4340445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786612.2 | XP_015642098.1 | 1 - 2 | 641 | 97 | |
| GenBank mRNA | AB101651.1 | - | 1 - 2 | 58 | 97 | |
More Information
Set Membership: |
|

LOC4340445
Genomic Map