Assay Details
| Gene Symbol: LOC4340605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787979.2 | XP_015643465.1 | 1 - 2 | 403 | 97 | |
| GenBank mRNA | AK102383.1 | - | 1 - 2 | 388 | 97 | |
More Information
Set Membership: |
|

LOC4340605
Genomic Map