Assay Details
| Gene Symbol: LOC4340605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787979.2 | XP_015643465.1 | 2 - 3 | 504 | 121 | |
| GenBank mRNA | AK102383.1 | - | 2 - 3 | 489 | 121 | |
More Information
Set Membership: |
|

LOC4340605
Genomic Map