Assay Details
| Gene Symbol: LOC4340605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787979.2 | XP_015643465.1 | 3 - 4 | 549 | 82 | |
| GenBank mRNA | AK102383.1 | - | 3 - 4 | 534 | 82 | |
More Information
Set Membership: |
|

LOC4340605
Genomic Map