Assay Details
| Gene Symbol: LOC4340605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787979.2 | XP_015643465.1 | 4 - 5 | 629 | 86 | |
| GenBank mRNA | AK102383.1 | - | 4 - 5 | 614 | 86 | |
More Information
Set Membership: |
|

LOC4340605
Genomic Map