Assay Details
| Gene Symbol: LOC4352284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK242914.1 | - | 2960 | 64 | ||
| Gene Symbol: LOC4338485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784467.2 | XP_015639953.1 | 2 - 3 | 215 | 64 | |
| GenBank mRNA | AF529175.1 | - | 2 - 3 | 178 | 64 | |
| AK061627.1 | - | 2 - 3 | 189 | 64 | ||
| AK103081.1 | - | 2 - 3 | 189 | 64 | ||
| AK104191.1 | - | 2 - 3 | 189 | 64 | ||
| BT129368.1 | - | 2 - 3 | 184 | 64 | ||
| BT132274.1 | - | 2 - 3 | 182 | 64 | ||
More Information
Set Membership: |
|

LOC4352284
Genomic Map