Assay Details
| Gene Symbol: LOC4338527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784721.2 | XP_015640207.1 | 7 - 8 | 1234 | 84 | |
| XM_015784722.2 | XP_015640208.1 | 7 - 8 | 1237 | 84 | ||
| GenBank mRNA | AK060514.1 | - | 7 - 8 | 1150 | 84 | |
More Information
Set Membership: |
|

LOC4338527
Genomic Map