Assay Details
| Gene Symbol: LOC4338545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783813.2 | XP_015639299.1 | 7 - 8 | 1066 | 60 | |
| GenBank mRNA | AK073358.1 | - | 7 - 8 | 1033 | 60 | |
| AK073379.1 | - | 7 - 8 | 1033 | 60 | ||
More Information
Set Membership: |
|

LOC4338545
Genomic Map