Assay Details
| Gene Symbol: LOC4338560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784542.1 | XP_015640028.1 | 12 - 13 | 1705 | 98 | |
| GenBank mRNA | AK071469.1 | - | 12 - 13 | 1641 | 98 | |
More Information
Set Membership: |
|

LOC4338560
Genomic Map