Assay Details
| Gene Symbol: LOC4339594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785119.2 | XP_015640605.1 | 5 - 6 | 1320 | 134 | |
| GenBank mRNA | AK072239.1 | - | 5 - 6 | 997 | 134 | |
More Information
Set Membership: |
|

LOC4339594
Genomic Map