Assay Details
| Gene Symbol: LOC4339689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783969.2 | XP_015639455.1 | 11 - 12 | 2908 | 66 | |
| GenBank mRNA | AK058719.1 | - | 11 - 12 | 1520 | 66 | |
More Information
Set Membership: |
|

LOC4339689
Genomic Map