Assay Details
| Gene Symbol: LOC4339689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783969.2 | XP_015639455.1 | 5 - 6 | 1919 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4339689| Gene Symbol: LOC4339689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783969.2 | XP_015639455.1 | 5 - 6 | 1919 | 126 | |
Set Membership: |
|