Assay Details
| Gene Symbol: LOC9271035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788654.2 | XP_015644140.1 | 13 - 14 | 2645 | 100 | |
| XM_015788655.2 | XP_015644141.1 | 13 - 14 | 2646 | 100 | ||
| GenBank mRNA | AK121044.1 | - | 13 - 14 | 2646 | 100 | |
More Information
Set Membership: |
|

LOC9271035
Genomic Map