Assay Details
| Gene Symbol: LOC9271035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788654.2 | XP_015644140.1 | 20 - 21 | 3285 | 68 | |
| GenBank mRNA | AK121044.1 | - | 18 - 19 | 4534 | 68 | |
More Information
Set Membership: |
|

LOC9271035
Genomic Map