Assay Details
| Gene Symbol: LOC4341806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786512.2 | XP_015641998.1 | 1 - 2 | 1268 | 97 | |
| GenBank mRNA | AK108697.1 | - | 1 - 2 | 531 | 97 | |
More Information
Set Membership: |
|

LOC4341806
Genomic Map