Assay Details
| Gene Symbol: LOC4341859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788997.2 | XP_015644483.1 | 2 - 3 | 461 | 128 | |
| XM_015788998.2 | XP_015644484.1 | 2 - 3 | 462 | 128 | ||
| GenBank mRNA | AK110977.1 | - | 2 - 3 | 430 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788997.2, XM_015788998.2 transcripts.
Set Membership: |
|

LOC4341859
Genomic Map