Assay Details
Gene Symbol: LOC4341924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787696.2 | XP_015643182.1 | 2 - 3 | 618 | 67 | |
GenBank mRNA | AK063936.1 | - | 2 - 3 | 659 | 67 | |
AK073594.1 | - | 2 - 3 | 625 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
