Assay Details
| Gene Symbol: LOC4341924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787696.2 | XP_015643182.1 | 4 - 5 | 1393 | 136 | |
| GenBank mRNA | AK063936.1 | - | 6 - 7 | 1221 | 136 | |
| AK073594.1 | - | 4 - 5 | 1400 | 136 | ||
More Information
Set Membership: |
|

LOC4341924
Genomic Map